Learn More
Medchemexpress LLC NCGC00138783 TFA | 99.4% | C28H27F4N7O3S | 1 ML
SDP
Catalog No. 5000353841
Encompass_Preferred
NCGC00138783 TFA is a selective inhibitor targeting the CD47/SIRPα axis. It directly blocks the interaction between CD47 and SIRPα axis.
  • Selective inhibitor
  • Targets CD47/SIRPα axis
  • IC50 of 50 μM
  • Blocks CD47 and SIRPα axis interaction
Catalog No. 50-003-53841 Supplier Medchemexpress LLC Supplier No. HY143224B10MM/1ML
NCGC00138783 TFA is a selective inhibitor targeting the CD47/SIRPα axis. It directly blocks the interaction between CD47 and SIRPα axis.
  • Selective inhibitor
  • Targets CD47/SIRPα axis
  • IC50 of 50 μM
  • Blocks CD47 and SIRPα axis interaction