Learn More
Medchemexpress LLC Ncgc00244536 | 2003260-55-5 | MFCD31382180 | 99.7% | 382.45 g/mol | C25H22N2O2 | 1 MG
SDP
Catalog No. 5000249262
Encompass_Preferred
NCGC00244536 is a small-molecule inhibitor of the histone demethylase KDM4B with a reported IC50 of 10 nM. Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies investigating KDM4B-dependent processes.
  • Potent KDM4B inhibition (IC50 10 nM).
  • High purity: 99.7%.
  • Solid, light yellow to green yellow appearance.
  • Molecular formula C25H22N2O2; molecular weight 382.45 g/mol.
  • Available in milligram pack sizes suitable for research use.
  • Suitable for biochemical and cellular research applications.
Catalog No. 50-002-49262 Supplier Medchemexpress LLC Supplier No. HY1017991MG
NCGC00244536 is a small-molecule inhibitor of the histone demethylase KDM4B with a reported IC50 of 10 nM. Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies investigating KDM4B-dependent processes.
  • Potent KDM4B inhibition (IC50 10 nM).
  • High purity: 99.7%.
  • Solid, light yellow to green yellow appearance.
  • Molecular formula C25H22N2O2; molecular weight 382.45 g/mol.
  • Available in milligram pack sizes suitable for research use.
  • Suitable for biochemical and cellular research applications.