Learn More
TARGETMOL CHEMICALS INC NCGC00244536 5MG
SDP
Catalog No. 502578840
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCGC00244536 (KDM4B Inhibitor B3) is a potent KDM4B inhibitor (IC50 10 nM). purity: 99%
Catalog No. 50-257-8840 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ00505MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCGC00244536 (KDM4B Inhibitor B3) is a potent KDM4B inhibitor (IC50 10 nM). purity: 99%