Learn More
Medchemexpress LLC HY-112308 5mg Medchemexpress, NCGC00247743 CAS:1435192-04-3 Purity:>98%
SDP
Catalog No. 501969079
Encompass_Preferred
Medchemexpress, HY-112308 5mg NCGC00247743 CAS:1435192-04-3 NCGC00247743 is a histone lysine demethylase KDM4 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9079 Supplier Medchemexpress LLC Supplier No. HY1123085MG
Medchemexpress, HY-112308 5mg NCGC00247743 CAS:1435192-04-3 NCGC00247743 is a histone lysine demethylase KDM4 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.