Learn More
Medchemexpress LLC HY-133512 5mg Medchemexpress, NCGC00249987 CAS:1384864-80-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1335125MG
Encompass_Preferred
Medchemexpress, HY-133512 5mg NCGC00249987 CAS:1384864-80-5 NCGC00249987 is a highly selective and allosteric Tyr phosphatase activity of Eya2 inhibitor with IC50s of 3 μM and 6.9 μM for Eya2 ED and MBP-Eya2 FL. NCGC00249987 specifically targets migration, invadopodia formation, and invasion of lung cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1343