Learn More
Abcam NCH 51, HDAC inhibitor, 50MG
Catalog No. 502707098
Encompass_Preferred
MW 390.6 Da, Purity >99%. Novel HDAC inhibitor (IC₅₀ values are 32, 48 and 41 nM at HDAC4, HDAC1 and HDAC6, respectively). Induces apoptosis and inhibits cell growth of various malignant cells and tumor cells lines (EC₅₀ = 3.8 μM). Also reactivates latent HIV-1 gene expression.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7098 Supplier Abcam Supplier No. AB12094950MG
MW 390.6 Da, Purity >99%. Novel HDAC inhibitor (IC₅₀ values are 32, 48 and 41 nM at HDAC4, HDAC1 and HDAC6, respectively). Induces apoptosis and inhibits cell growth of various malignant cells and tumor cells lines (EC₅₀ = 3.8 μM). Also reactivates latent HIV-1 gene expression.
The product is subject to the following: Abcam Restricted Use Statement