Learn More
FENICSBIO CORP NCI-H929/GFP STABLE CELL LINE
Supplier:  FENICSBIO CORP CL1643
Encompass
NCI-H929/GFP cells are derived from the human B lymphocyte NCI-H929 cell line by stably integration of a constitutive GFP stably expression construct. NCI-H929 cells stably express B cell maturation antigen (BCMA)-a novel treatment target for multiple myeloma (MM), and have been widely used in cancer research and drug development. NCI-H929/GFP cells stably stably expressGFP, can be used for in vitro assays and in vivo imaging.
Catalog No. NC3385360