Learn More
Medchemexpress LLC HY-U00155 1mg Medchemexpress, NCI172112 CAS:56605-16-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001551MG
Encompass_Preferred
Medchemexpress, HY-U00155 1mg NCI172112 CAS:56605-16-4 NCI172112 is a classical bifunctional alkylating agent synthesized in an effort to develop antitumor agents effective against CNS tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3189