Learn More
ATCC NCI-BL2171; EBV-transformed cells; Human (Homo sapiens)
Catalog No. 502383829
Encompass
NCI-BL2171 [BL2171] are B lymphoblasts that were isolated from the peripheral blood of a normal 50-year-old, White male donor. This product has applications in immunology.
Catalog No. 50-238-3829 Supplier ATCC Supplier No. CRL5969
NCI-BL2171 [BL2171] are B lymphoblasts that were isolated from the peripheral blood of a normal 50-year-old, White male donor. This product has applications in immunology.