Learn More
ATCC NCI-H1155; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382288
Encompass
NCI-H1155 [H1155] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 36-year-old, White male with carcinoma. This product has applications for cancer research.
Catalog No. 50-238-2288 Supplier ATCC Supplier No. CRL5818
NCI-H1155 [H1155] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 36-year-old, White male with carcinoma. This product has applications for cancer research.