Learn More
ATCC NCI-H1299; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502381692
Encompass
NCI-H1299 is an epithelial-like cell that was isolated from the lung of a White, 43-year-old, male patient with carcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in cancer and immuno-oncology research.
Catalog No. 50-238-1692 Supplier ATCC Supplier No. CRL5803
NCI-H1299 is an epithelial-like cell that was isolated from the lung of a White, 43-year-old, male patient with carcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in cancer and immuno-oncology research.