Learn More
ATCC NCI-H1341; Lung Cancer; Human (Homo sapiens)
Catalog No. 502382280
Encompass
NCI-H1341 [H1341] is a cell line that was isolated from the lungs of a non-smoker female with carcinoma. This product can be used for cancer research.
Catalog No. 50-238-2280 Supplier ATCC Supplier No. CRL5864
NCI-H1341 [H1341] is a cell line that was isolated from the lungs of a non-smoker female with carcinoma. This product can be used for cancer research.