Learn More
ATCC NCI-H1395; Lung Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5868
Encompass
NCI-H1395 [H1395] is a cell line exhibiting epithelial-like morphology that was isolated from a 55-year-old, White female with adenocarcinoma. This product can be used for cancer research.
Catalog No. 50-238-4154