Learn More
ATCC NCI-H146; Small Cell Lung Cancer; Human (Homo sapiens)
Catalog No. 502383848
Encompass
NCI-H146 [H146] is an epithelial-like cell that was isolated from the lung of a 59-year-old, White, male with lung carcinoma. The cell line NCI-H146 [H146] can be used in cancer and immunology research.
Catalog No. 50-238-3848 Supplier ATCC Supplier No. HTB173
NCI-H146 [H146] is an epithelial-like cell that was isolated from the lung of a 59-year-old, White, male with lung carcinoma. The cell line NCI-H146 [H146] can be used in cancer and immunology research.