Learn More
ATCC NCI-H1568; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384146
Encompass
NCI-H1568 [H1568] cells were isolated from the lungs of a 48-year-old, White female patient with non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-4146 Supplier ATCC Supplier No. CRL5876
NCI-H1568 [H1568] cells were isolated from the lungs of a 48-year-old, White female patient with non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.