Learn More
ATCC NCI-H1648; Lung Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5882
Encompass
NCI-H1648 [H1648] are lymphoblasts exhibiting epithelial morphology that was isolated from the lungs of a 39-year-old, Black male with stage 3A adenocarcinoma. This product has applications in immunology.
Catalog No. 50-238-4754