Learn More
ATCC NCI-H1666; Lung Carcinoma; Human
Catalog No. 502384155
Encompass
NCI-H1666 [H-1666, H1666] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 50-year-old, White female patient with bronchoalveolar carcinoma. This product has applications in cancer research.
Catalog No. 50-238-4155 Supplier ATCC Supplier No. CRL5885
Add to Cart
Add to Cart
NCI-H1666 [H-1666, H1666] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 50-year-old, White female patient with bronchoalveolar carcinoma. This product has applications in cancer research.