Learn More
ATCC NCI-H1693; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382861
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
NCI-H1693 [H1693] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 55-year-old, White female patient with stage 3B non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-2861 Supplier ATCC Supplier No. CRL5887
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
NCI-H1693 [H1693] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 55-year-old, White female patient with stage 3B non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.