Learn More
ATCC NCI-H1755; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382489
Encompass
NCI-H1755 [H1755] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 65-year-old White, female patient with non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-2489 Supplier ATCC Supplier No. CRL5892
NCI-H1755 [H1755] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 65-year-old White, female patient with non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.