Learn More
ATCC NCI-H1781; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383026
Encompass
NCI-H1781 [H1781] is a cell line isolated from the lungs of a 66-year-old, White female with stage 3B bronchoalveolar carcinoma. This product has applications in cancer research.
Catalog No. 50-238-3026 Supplier ATCC Supplier No. CRL5894
 Restricted Item
Add to Cart
Add to Cart
NCI-H1781 [H1781] is a cell line isolated from the lungs of a 66-year-old, White female with stage 3B bronchoalveolar carcinoma. This product has applications in cancer research.