Learn More
ATCC NCI-H1838; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502382340
Encompass
NCI-H1838 [H1838] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a female patient with non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-2340 Supplier ATCC Supplier No. CRL5899
NCI-H1838 [H1838] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a female patient with non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.