Learn More
ATCC NCI-H1963; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383412
Encompass
NCI-H1963 [H1963] is a cell line that was isolated from the lungs of a 56-year-old, Black male patient with stage L small cell lung cancer: carcinoma. This product has applications in cancer research.
Catalog No. 50-238-3412 Supplier ATCC Supplier No. CRL5982
Add to Cart
Add to Cart
NCI-H1963 [H1963] is a cell line that was isolated from the lungs of a 56-year-old, Black male patient with stage L small cell lung cancer: carcinoma. This product has applications in cancer research.