Learn More
ATCC NCI-H1975; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382785
Encompass
NCI-H1975 [H-1975, H1975] is a cell line exhibiting epithelial morphology that was isolated in 1988 from the lungs of a nonsmoking female with non-small cell lung cancer. Use these cells in immuno-oncology and lung cancer research.
Catalog No. 50-238-2785 Supplier ATCC Supplier No. CRL5908
NCI-H1975 [H-1975, H1975] is a cell line exhibiting epithelial morphology that was isolated in 1988 from the lungs of a nonsmoking female with non-small cell lung cancer. Use these cells in immuno-oncology and lung cancer research.