Learn More
ATCC NCI-H1975 Quantitative; purified DNA, 3 ug; Human
Supplier:  ATCC CRL5908DQ
Encompass
Quantified for oncology biomarker EGFR p.T790M and EGFR p.L858R via NGS and ddPCR
Catalog No. 50-238-2393