Learn More
ATCC NCI-H1993; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502382634
Encompass
NCI-H1993 [H1993] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 47-year-old, White female patient with stage 3A non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-2634 Supplier ATCC Supplier No. CRL5909
NCI-H1993 [H1993] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 47-year-old, White female patient with stage 3A non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.