Learn More
ATCC NCI-H2023; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502382134
Encompass
NCI-H2023 [H2023] is a cell line that was isolated from the lungs of a 26-year-old, White, male patient with stage 3A non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-2134 Supplier ATCC Supplier No. CRL5912
NCI-H2023 [H2023] is a cell line that was isolated from the lungs of a 26-year-old, White, male patient with stage 3A non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.