Learn More
ATCC NCI-H2029; Lung Carcinoma; Human
Catalog No. 502381859
Encompass
NCI-H2029 [H2029] is a cell line that was isolated from the lungs of a 69-year-old, White, female patient with stage E small cell lung cancer: carcinoma. This product has applications in cancer research.
Catalog No. 50-238-1859 Supplier ATCC Supplier No. CRL5913
NCI-H2029 [H2029] is a cell line that was isolated from the lungs of a 69-year-old, White, female patient with stage E small cell lung cancer: carcinoma. This product has applications in cancer research.