Learn More
ATCC NCI-H2052; Mesothelioma; Human (Homo sapiens)
Supplier:  ATCC CRL5915
Encompass
NCI-H2052 [H2052] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 65-year-old, White, male patient with stage 4 mesothelioma. This product has applications in cancer research.
Catalog No. 50-238-4846