Learn More
ATCC NCI-H2073; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383095
Encompass
NCI-H2073 [H2073] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 47-year-old, White, female patient with stage 3A non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-3095 Supplier ATCC Supplier No. CRL5918
NCI-H2073 [H2073] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 47-year-old, White, female patient with stage 3A non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.