Learn More
ATCC NCI-H209; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB172
Encompass
NCI-H209 [H209] are round cells that were isolated from the bone marrow of a White male with lung carcinoma. The bone marrow specimen was take prior to therapy. NCI-H209 [H209] can be used in cancer and immunology research.
Catalog No. 50-238-3865