Learn More
ATCC NCI-H2122; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384304
Encompass
NCI-H2122 [H2122] cells are lymphoblasts isolated in 1989 from a pleural effusion metastasis derived from a 46-year-old female smoker. Use these cells in lung cancer research.
Catalog No. 50-238-4304 Supplier ATCC Supplier No. CRL5985
NCI-H2122 [H2122] cells are lymphoblasts isolated in 1989 from a pleural effusion metastasis derived from a 46-year-old female smoker. Use these cells in lung cancer research.