Learn More
ATCC NCI-H2171; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383681
Encompass
NCI-H2171 [H2171] are lymphoblasts exhibiting floating clusters morphology that was isolated from the lungs of a 50-year-old, White male patient with stage E small cell lung cancer: carcinoma. This product has applications in immunology and cancer research.
Catalog No. 50-238-3681 Supplier ATCC Supplier No. CRL5929
NCI-H2171 [H2171] are lymphoblasts exhibiting floating clusters morphology that was isolated from the lungs of a 50-year-old, White male patient with stage E small cell lung cancer: carcinoma. This product has applications in immunology and cancer research.