Learn More
ATCC NCI-H2227; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502384779
Encompass
NCI-H2227 [H2227] is a cell line that was isolated from the lungs of a 54-year-old, white male patient with stage E small cell lung cancer: carcinoma. This product has applications in cancer research.
Catalog No. 50-238-4779 Supplier ATCC Supplier No. CRL5934
NCI-H2227 [H2227] is a cell line that was isolated from the lungs of a 54-year-old, white male patient with stage E small cell lung cancer: carcinoma. This product has applications in cancer research.