Learn More
ATCC NCI-H2286; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383857
Encompass
NCI-H2286 [H2286] is a cell line exhibiting rounded morphology that was isolated from the lungs of a 57-year-old, White female patient with stage 1 small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-3857 Supplier ATCC Supplier No. CRL5938
NCI-H2286 [H2286] is a cell line exhibiting rounded morphology that was isolated from the lungs of a 57-year-old, White female patient with stage 1 small cell lung cancer: adenocarcinoma. This product has applications in cancer research.