Learn More
ATCC NCI-H2291; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383861
Encompass
NCI-H2291 [H2291] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a male patient with small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-3861 Supplier ATCC Supplier No. CRL5939
NCI-H2291 [H2291] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a male patient with small cell lung cancer: adenocarcinoma. This product has applications in cancer research.