Learn More
ATCC NCI-H23; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382330
Encompass
NCI-H23 [H23] is an epithelial-like cell that was isolated from the lung of a Black, 51-year-old, male patient with adenocarcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in toxicology and cancer research.
Catalog No. 50-238-2330 Supplier ATCC Supplier No. CRL5800
 Restricted Item
Add to Cart
Add to Cart
NCI-H23 [H23] is an epithelial-like cell that was isolated from the lung of a Black, 51-year-old, male patient with adenocarcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in toxicology and cancer research.