Learn More
ATCC NCI-H28; Mesothelioma; Human (Homo sapiens)
Supplier:  ATCC CRL5820
Encompass
NCI-H28 [H28] is a cell line that was isolated from the lungs of a 48-year-old, White male with stage 4 mesothelioma. This product has applications for cancer research.
Catalog No. 50-238-4423
 Restricted Item