Learn More
ATCC NCI-H295R; Adrenal Carcinoma; Human (Homo sapiens)
Catalog No. 502383237
Encompass
NCI-H295R [H295R] are cells that were isolated from the adrenal gland of a 48-year-old, Black, female adrenal cancer patient. This cell line has the ability to produce adrenal androgens.
Catalog No. 50-238-3237 Supplier ATCC Supplier No. CRL2128
NCI-H295R [H295R] are cells that were isolated from the adrenal gland of a 48-year-old, Black, female adrenal cancer patient. This cell line has the ability to produce adrenal androgens.