Learn More
ATCC NCI-H358; Bronchioalveolar Carcinoma; Human (Homo sapiens)
Catalog No. 502384463
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
NCI-H358 [H-358, H358] is an epithelial-like cell that was isolated from the bronchiole of a male patient with bronchioalveolar carcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in cancer research.
Catalog No. 50-238-4463 Supplier ATCC Supplier No. CRL5807
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
NCI-H358 [H-358, H358] is an epithelial-like cell that was isolated from the bronchiole of a male patient with bronchioalveolar carcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in cancer research.