Learn More
ATCC NCI-H441; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383348
Encompass
NCI-H441 [H441] cells were isolated in 1982 from the pericardial fluid of a male patient with papillary adenocarcinoma of the lung. Use these cells as a transfection host for expression of pulmonary surfactant protein (SP-B).
Catalog No. 50-238-3348 Supplier ATCC Supplier No. HTB174
NCI-H441 [H441] cells were isolated in 1982 from the pericardial fluid of a male patient with papillary adenocarcinoma of the lung. Use these cells as a transfection host for expression of pulmonary surfactant protein (SP-B).