Learn More
ATCC NCI-H446; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB171
Encompass
NCI-H446 [H446] cells were isolated in 1982 from the pleural fluid of a 61-year-old, White, male patient with small cell cancer of the lung.
Catalog No. 50-238-4758