Learn More
ATCC NCI-H508; Cecum Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383453
Encompass
NCI-H508 [H508] is a cell line derived from metastasis to the abdominal wall obtained from a patient after treatment with 5-fluorouracil. NCI-H508 [H508] is from the cecum of a White, 55-year-old, male patient with colorectal adenocarcinoma. This cell line was deposited by AF Gazdar.
Catalog No. 50-238-3453 Supplier ATCC Supplier No. CCL253
NCI-H508 [H508] is a cell line derived from metastasis to the abdominal wall obtained from a patient after treatment with 5-fluorouracil. NCI-H508 [H508] is from the cecum of a White, 55-year-old, male patient with colorectal adenocarcinoma. This cell line was deposited by AF Gazdar.