Learn More
ATCC NCI-H510A; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB184
Encompass
NCI-H510A [H510A, NCI-H510] is an epithelial-like cell that was isolated from an adrenal metastasis of a 56-year-old, White, male with lung carcinoma.
Catalog No. 50-238-3545