Learn More
ATCC NCI-H522; Lung Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5810
Encompass
NCI-H522 [H522] is a cell line exhibiting epithelial morphology that were isolated from the lungs of a 58-year-old White male with adenocarcinoma. This product has applications in cancer research and toxicology.
Catalog No. 50-238-4483