Learn More
ATCC NCI-H647; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5834
Encompass
NCI-H647 [H647] is a cell line that was isolated from the lungs of a 56-year-old, White male with stage 3A adenosquamous carcinoma. This product has applications for cancer research.
Catalog No. 50-238-4450