Learn More
ATCC NCI-H660; Prostate Carcinoma; Human (Homo sapiens)
Catalog No. 502382728
Encompass
NCI-H660 is an epithelial neuroendocrine cell that was isolated from the prostate of a White, 63-year-old, male patient with carcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in cancer research.
Catalog No. 50-238-2728 Supplier ATCC Supplier No. CRL5813
NCI-H660 is an epithelial neuroendocrine cell that was isolated from the prostate of a White, 63-year-old, male patient with carcinoma. This cell line was deposited by AF Gazdar and JD Minna and can be used in cancer research.