Learn More
ATCC NCI-H661; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB183
Encompass
NCI-H661 [H661] is an epithelial-like cell that was isolated from the lung of a 43-year-old, White, male with lung carcinoma. The cell line can be used in cancer research.
Catalog No. 50-238-2471