Learn More
ATCC NCI-H889; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383084
Encompass
NCI-H889 [H889] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 69-year-old, White female with carcinoma. This product has applications for cancer research.
Catalog No. 50-238-3084 Supplier ATCC Supplier No. CRL5817
NCI-H889 [H889] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 69-year-old, White female with carcinoma. This product has applications for cancer research.