Learn More
ATCC NCI-H929; Plasmacytoma; Human (Homo sapiens)
Catalog No. 502382612
Encompass
NCI-H929 [H929] cells are B lymphocytes isolated from a malignant effusion in a 62-year-old, White, female plasmacytoma myeloma patient. Use these cells in cancer and immunology research. This product is an ATCC manufactured and accessioned progeny of ATCC CRL-9068 cited in US Pat. No. 4,892,829.
Catalog No. 50-238-2612 Supplier ATCC Supplier No. CRL3580
Add to Cart
Add to Cart
NCI-H929 [H929] cells are B lymphocytes isolated from a malignant effusion in a 62-year-old, White, female plasmacytoma myeloma patient. Use these cells in cancer and immunology research. This product is an ATCC manufactured and accessioned progeny of ATCC CRL-9068 cited in US Pat. No. 4,892,829.