Learn More
New England Biolabs, Inc. NciI – 2000 units
SDP
Catalog No. 50812129
Encompass
A restriction endonuclease that recognizes the sequence CC^S_GG.
Catalog No. 50-812-129 Supplier New England Biolabs, Inc. Supplier No. R0196S
A restriction endonuclease that recognizes the sequence CC^S_GG.